Tisk…
2,5-Dimethyl-1,5-hexadien-3-ol ≥97%
Dodavatel: Thermo Fisher Scientific
B20072.14 B20072.06
B20072.14EA 5260 CZK
B20072.14 B20072.06
2,5-Dimethyl-1,5-hexadien-3-ol ≥97%
2,5-Dimethyl-1,5-hexadien-3-ol
Purity: 97% CAS No.: 17123-63-6 Molecular Formula: C8H14O Formula Weight: 126.20 MDL No.: MFCD00026056
Vzorec: C₈H₁₄O
MW: 126.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026056
CAS číslo: 17123-63-6