Tisk…

Methyl trifluoropyruvate ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Methyl 3,3,3-trifluoropyruvate, Methyl 3,3,3-trifluoro-2-oxopropanoate

B21589.06 B21589.14
B21589.06EA 2440 CZK
B21589.06 B21589.14
Methyl trifluoropyruvate ≥97%
Methyl trifluoropyruvate

Purity: 97% CAS No.: 13089-11-7 Molecular Formula: C4H3F3O3 Formula Weight: 156.06 Flash Point: 26°(79°F) Refractive Index: 1.3320 Boiling Point: 84-86° Beilstein Reference: 1863490 UN No.: 3272 MDL No.: MFCD00114936

Vzorec: C₄H₃F₃O₃
MW: 156,06 g/mol
Bod varu: 84…86 °C
Hustota: 1,52
Bod vzplanutí: 26 °C (79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00114936
CAS číslo: 13089-11-7
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR