3-Fluoro-4-pyridineboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(3-Fluoropyridin-4-yl)boronic acid
H53109.MD
H53109.03
H53109.MDEA
1900
CZK
H53109.MD
H53109.03
3-Fluoro-4-pyridineboronic acid ≥98%
3-Fluoro-4-pyridineboronic acid
Purity: 98%
CAS No.: 458532-97-3
Molecular Formula: C5H5BFNO2
Formula Weight: 140.91
Melting Point: 237-239°
MDL No.: MFCD03788558
Vzorec:
C₅H₅BFNO₂ MW: 140.91 g/mol Bod tání: 237…239 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788558 CAS číslo: 458532-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...