Tisk…

2-Propylthiazole ≥99%

Dodavatel: Thermo Scientific
Varování

A11574.06 A11574.14
A11574.06EA 2510 CZK
A11574.06 A11574.14
2-Propylthiazole ≥99%
2-Propylthiazole

Purity: 99% CAS No.: 17626-75-4 Molecular Formula: C6H9NS Formula Weight: 127.20 Flash Point: 54°(129°F) Refractive Index: 1.5055 Boiling Point: 62°/15mm UN No.: 1993 MDL No.: MFCD00053154

Vzorec: C₆H₉NS
MW: 127,20 g/mol
Bod varu: 62 °C (15 mmHg)
Hustota: 1,03
Bod vzplanutí: 54 °C (129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053154
CAS číslo: 17626-75-4
EINECS: 241-606-7
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR