Tisk…

tert-Butyl 2-bromoisobutyrate 97%

Dodavatel: Thermo Scientific
Nebezpečí

L08216.18 L08216.30 L08216.09
L08216.18EA 2030 CZK
L08216.18 L08216.30 L08216.09
tert-Butyl 2-bromoisobutyrate 97%
tert-Butyl 2-bromoisobutyrate

Purity: 97% CAS No.: 23877-12-5 Molecular Formula: C8H15BrO2 Formula Weight: 223.12 Storage Temperature: 2°C to 8°C Refractive Index: 1.4360 Boiling Point: 35-37°/1mm Beilstein Reference: 1905654 MDL No.: MFCD00051908

Vzorec: C₆H₁₁BrO₂
Bod varu: 35…37 °C (1 mmHg)
Hustota: 1,180
Storage Temperature: Refrigerator
MDL Number: MFCD00051908
CAS číslo: 23877-12-5
EINECS: 245-924-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR