Tisk…

2-Chloro-6-methylbenzonitrile ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

L00706.03 L00706.06
L00706.03EA 961 CZK
L00706.03 L00706.06
2-Chloro-6-methylbenzonitrile ≥98%
2-Chloro-6-methylbenzonitrile

Purity: 98% CAS No.: 6575-09-3 Molecular Formula: C8H6ClN Formula Weight: 151.60 Odour: Characteristic Melting Point: 79-83° Boiling Point: 143°/38mm Beilstein Reference: 2246414 UN No.: 3439 MDL No.: MFCD00001782

Vzorec: C₈H₆ClN
MW: 151,60 g/mol
Bod varu: 143 °C (38 mmHg)
Bod tání: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00001782
CAS číslo: 6575-09-3
EINECS: 229-499-5
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR