Tisk…

4-(Chloromethyl)benzonitrile ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B22138.09 B22138.18 B22138.30
B22138.09EA 910 CZK
B22138.09 B22138.18 B22138.30
4-(Chloromethyl)benzonitrile ≥98%
4-(Chloromethyl)benzonitrile

Purity: 98+% CAS No.: 874-86-2 Molecular Formula: C8H6ClN Formula Weight: 151.60 Melting Point: 77-80° UN No.: 2923 MDL No.: MFCD00019761

Vzorec: C₈H₆ClN
Bod tání: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00019761
CAS číslo: 874-86-2
EINECS: 212-869-5
UN: 2923
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR