5,6,7,8-Tetrahydroquinoxaline ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L11204.03
L11204.06
L11204.03EA
920
CZK
L11204.03
L11204.06
5,6,7,8-Tetrahydroquinoxaline ≥99%
5,6,7,8-Tetrahydroquinoxaline
Purity: 99%
CAS No.: 34413-35-9
Molecular Formula: C8H10N2
Formula Weight: 134.18
Flash Point: 92°(197°F)
Refractive Index: 1.5420
Boiling Point: 72-74°/4mm
Beilstein Reference: 957053
MDL No.: MFCD00038078
Vzorec:
C₈H₁₀N₂ Bod varu: 72…74 °C (4 mmHg) Hustota: 1,061 Bod vzplanutí: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00038078 CAS číslo: 34413-35-9 EINECS: 252-002-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...