Tisk…
Boron trichloride 1 mol/l in p-xylene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
345458-100ML 345458-800ML
SIAL345458-100MLEA 1750 CZK
SIAL345458-100ML SIAL345458-800ML
Boron trichloride 1 mol/l in p-xylene, Sigma-Aldrich®
Boron trichloride
Vzorec: BCl₃
MW: 117.17
Bod varu: 12.4
Bod tání: -107
Hustota: 1.876
Storage Temperature: Ambient
MDL Number: MFCD00011313
CAS číslo: 10294-34-5
EINECS: 233-658-4
Merck Index: 12,01378