Tisk…

(±)-tert-Butyl-2-bromobutyrate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: tert-Butyl 2-bromobutanoate, tert-Butyl 2-bromobutyrate

L13799.06 L13799.14
L13799.06EA 1050 CZK
L13799.06 L13799.14
(±)-tert-Butyl-2-bromobutyrate ≥98%
(±)-tert-Butyl-2-bromobutyrate

Purity: 98% CAS No.: 24457-21-4 Molecular Formula: C8H15BrO2 Formula Weight: 223.12 Refractive Index: 1.4410 Boiling Point: 72-76°/12mm UN No.: 1760 MDL No.: MFCD00209634

Vzorec: C₈H₁₅BrO₂
Bod varu: 72…76 °C (12 mmHg)
Hustota: 1,200
Storage Temperature: Ambient
MDL Number: MFCD00209634
CAS číslo: 24457-21-4
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR