Iodomethane[¹³C,D₃] (99% ¹³C , 99.5% D) stabilizovaný, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Iodomethane-[¹³C,²H₃]
Stabilization:
Stabilised with copper
M-Clarity™ quality level = MQ200
294756-1G
294756-5G
SIAL294756-1GEA
9060
CZK
SIAL294756-1G
SIAL294756-5G
Iodomethane[¹³C,D₃] (99% ¹³C , 99.5% D) stabilizovaný, Sigma-Aldrich®
Iodomethane[¹³C,D₃]
Vzorec:
CD₃I |
MDL Number:
MFCD00001076 CAS číslo: 20710-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...