Tisk…

(4-Methoxyphenyl)magnesium bromide 0.5 mol/l in THF, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Anisylmagnesium bromide

M-Clarity™ quality level = MQ100

470260-100ML 470260-1L
SIAL470260-100MLEA 2940 CZK
SIAL470260-100ML SIAL470260-1L
(4-Methoxyphenyl)magnesium bromide 0.5 mol/l in THF, Sigma-Aldrich®
(4-Methoxyphenyl)magnesium bromide
Vzorec: C₇H₇BrMgO
MW: 211,34 g/mol
MDL Number: MFCD00013515
CAS číslo: 13139-86-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR