2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
H54793.MD
H54793.03
H54793.MDEA
3540
CZK
H54793.MD
H54793.03
2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 96%
2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester
Purity: 96%
CAS No.: 947249-01-6
Molecular Formula: C12H16BF3N2O2
Formula Weight: 288.07
Melting Point: 132-134°
MDL No.: MFCD12923414
Vzorec:
C₁₂H₁₆BF₃N₂O₂ MW: 288,07 g/mol Bod tání: 132…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD12923414 CAS číslo: 947249-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...