Zinc titanate ≥99.9% (metals basis)
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Zinc titanium oxide
41673.18
41673.30
41673.18EA
2970
CZK
41673.18
41673.30
Zinc titanate ≥99.9% (metals basis)
Zinc titanate
Purity: 99.9%
CAS No.: 12036-69-0
Molecular Formula: Zn2TiO4, may contain small amounts of ZnO and TiO2
Formula Weight: 242.64
Physical Form: -200 Mesh Powder
Odour: Odourless
MDL No.: MFCD00054107
Vzorec:
O₃TiZn MW: 242,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00054107 CAS číslo: 12036-43-0 EINECS: 234-850-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...