3-Amino-1-bromoisoquinoline ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Bromoisoquinolin-3-amine
H50368.03
H50368.06
H50368.03EA
5940
CZK
H50368.03
H50368.06
3-Amino-1-bromoisoquinoline ≥97%
3-Amino-1-bromoisoquinoline
Purity: 97+%
CAS No.: 13130-79-5
Molecular Formula: C9H7BrN2
Formula Weight: 223.07
MDL No.: MFCD00102193
Vzorec:
C₉H₇BrN₂ MW: 223,07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00102193 CAS číslo: 13130-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...