Tisk…

5-Bromo-8-methoxyquinoline 96%

Dodavatel: Thermo Scientific
Varování

H54386.MD H54386.03 H54386.06
H54386.MDEA 1460 CZK
H54386.MD H54386.03 H54386.06
5-Bromo-8-methoxyquinoline 96%
5-Bromo-8-methoxyquinoline

Purity: 96% CAS No.: 10522-47-1 Molecular Formula: C10H8BrNO Formula Weight: 238.08 Storage Temperature: 2°C to 8°C Melting Point: 80-82° MDL No.: MFCD04966996

Vzorec: C₁₀H₈BrNO
MW: 238,08 g/mol
Bod tání: 80…82 °C
MDL Number: MFCD04966996
CAS číslo: 10522-47-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR