Tisk…

6-Bromo-2-chloroquinoline 96%

Dodavatel: Thermo Scientific
Varování

H54895.03 H54895.06
H54895.03EA 4570 CZK
H54895.03 H54895.06
6-Bromo-2-chloroquinoline 96%
6-Bromo-2-chloroquinoline

Purity: 96% CAS No.: 1810-71-5 Molecular Formula: C9H5BrClN Formula Weight: 242.50 Storage Temperature: 2°C to 8°C Melting Point: 148-150° MDL No.: MFCD04115272

Vzorec: C₉H₅BrClN
MW: 242,50 g/mol
Bod tání: 148…150 °C
MDL Number: MFCD04115272
CAS číslo: 1810-71-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR