Tisk…

8-Quinolineboronic acid pinacol ester 95%

Dodavatel: Thermo Fisher Scientific

H51698.03 H51698.06
H51698.03EA 2320 CZK
H51698.03 H51698.06
8-Quinolineboronic acid pinacol ester 95%
8-Quinolineboronic acid pinacol ester

Purity: 95% CAS No.: 190788-62-6 Molecular Formula: C15H18BNO2 Formula Weight: 255.12 MDL No.: MFCD05663894

Vzorec: C₁₅H₁₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD05663894
CAS číslo: 190788-62-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR