2-(Benzyloxy)phenylboronic acid 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Benzyloxyphenylboronic acid, (2-(Benzyloxy)phenyl)boronic acid, 2-Benzyloxybenzeneboronic acid
L20100.03
L20100.06
L20100.03EA
1590
CZK
L20100.03
L20100.06
2-(Benzyloxy)phenylboronic acid 96%
2-(Benzyloxy)phenylboronic acid
Purity: 96%
CAS No.: 190661-29-1
Molecular Formula: C13H13BO3
Formula Weight: 228.06
Melting Point: 96-99°
Beilstein Reference: 7813704
Vzorec:
C₁₃H₁₃BO₃ MW: 228.06 g/mol Bod tání: 96…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632206 CAS číslo: 190661-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...