Methyl-3-iodo-4-methylbenzoate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Methyl 3-iodo-4-methylbenzoate
H28734.14
H28734.06
H28734.14EA
4770
CZK
H28734.14
H28734.06
Methyl-3-iodo-4-methylbenzoate ≥98%
Methyl-3-iodo-4-methylbenzoate
Purity: 98%
CAS No.: 90347-66-3
Molecular Formula: C9H9IO2
Formula Weight: 276.07
Refractive Index: 1.5975
Vzorec:
C₉H₉IO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00230583 CAS číslo: 90347-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...