Tisk…

Methyl-3-methoxy-4-methylbenzoate ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Methyl 3-methoxy-4-methylbenzoate

B22695.06 B22695.14
B22695.06EA 1720 CZK
B22695.06 B22695.14
Methyl-3-methoxy-4-methylbenzoate ≥98%
Methyl-3-methoxy-4-methylbenzoate

Purity: 98% CAS No.: 3556-83-0 Molecular Formula: C10H12O3 Formula Weight: 180.21 Flash Point: 110°(230°F) Refractive Index: 1.5295 Melting Point: 47-50° Boiling Point: 119-120°/1mm Beilstein Reference: 1948339 MDL No.: MFCD00082710

Vzorec: C₁₀H₁₂O₃
Bod varu: 119…120 °C (1 mmHg)
Bod tání: 47…50 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082710
CAS číslo: 3556-83-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR