Tisk…

4-Bromo-3-methylbenzamide ≥99%

Dodavatel: Thermo Scientific

L16176.14 L16176.03 L16176.06
L16176.14EA 5010 CZK
L16176.14 L16176.03 L16176.06
4-Bromo-3-methylbenzamide ≥99%
4-Bromo-3-methylbenzamide

Purity: 99% CAS No.: 170229-98-8 Molecular Formula: C8H8BrNO Formula Weight: 214.06 Melting Point: 177-182° Beilstein Reference: 868074

Vzorec: C₈H₈BrNO
MW: 214,06 g/mol
Bod tání: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00672914
CAS číslo: 170229-98-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR