4-Methoxybenzenesulphonamide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L08961.06
L08961.14
L08961.06EA
2280
CZK
L08961.06
L08961.14
4-Methoxybenzenesulphonamide ≥98%
4-Methoxybenzenesulphonamide
Purity: 98%
CAS No.: 1129-26-6
Molecular Formula: C7H9NO3S
Formula Weight: 187.22
Melting Point: 110-114°
Beilstein Reference: 2047881
Vzorec:
C₇H₉NO₃S MW: 187.22 g/mol Bod tání: 110…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025392 CAS číslo: 1129-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...