N-Cyclopropyl-4-nitrobenzenesulphonamide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H58673.MC
H58673.ME
H58673.MCEA
1930
CZK
H58673.MC
H58673.ME
N-Cyclopropyl-4-nitrobenzenesulphonamide ≥97%
N-Cyclopropyl-4-nitrobenzenesulphonamide
Purity: 97%
CAS No.: 549476-61-1
Molecular Formula: C9H10N2O4S
Formula Weight: 242.25
MDL No.: MFCD00458256
Vzorec:
C₉H₁₀N₂O₄S Storage Temperature: Ambient |
MDL Number:
MFCD00458256 CAS číslo: 549476-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...