4-Fluoro-3-nitrobenzenesulphonamide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H54516.MD
H54516.03
H54516.MDEA
2290
CZK
H54516.MD
H54516.03
4-Fluoro-3-nitrobenzenesulphonamide ≥97%
4-Fluoro-3-nitrobenzenesulphonamide
Purity: 97%
CAS No.: 406233-31-6
Molecular Formula: C6H5FN2O4S
Formula Weight: 220.18
MDL No.: MFCD08703185
Vzorec:
C₆H₅FN₂O₄S MW: 220.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08703185 CAS číslo: 406233-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...