Tisk…

2-Fluoro-N-methylbenzamide ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H56104.MD H56104.03
H56104.MDEA 2810 CZK
H56104.MD H56104.03
2-Fluoro-N-methylbenzamide ≥97%
2-Fluoro-N-methylbenzamide

Purity: 97% CAS No.: 52833-63-3 Molecular Formula: C8H8FNO Formula Weight: 153.16 MDL No.: MFCD01214098

Vzorec: C₈H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD01214098
CAS číslo: 52833-63-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR