Tisk…

3-Amino-4-hydroxybenzonitrile 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-Amino-4-hydroxybenzonitrile, 2-Amino-4-cyanophenol, 2-Amino-4-cyano-phenol

H33856.03 H33856.06
H33856.03EA 6010 CZK
H33856.03 H33856.06
3-Amino-4-hydroxybenzonitrile 97%
3-Amino-4-hydroxybenzonitrile
Vzorec: C₇H₆N₂O
MW: 134,14 g/mol
Bod tání: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00234273
CAS číslo: 14543-43-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR