Tisk…

3-(Methylcarbamoyl)phenylboronic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-(N-Methylaminocarbonyl)phenylboronic acid, 3-(Methylcarbamoyl)benzeneboronic acid, (3-(Methylcarbamoyl)phenyl)boronic acid

H53141.MD H53141.03
H53141.MDEA 2420 CZK
H53141.MD H53141.03
3-(Methylcarbamoyl)phenylboronic acid ≥98%
3-(Methylcarbamoyl)phenylboronic acid

Purity: 98% CAS No.: 832695-88-2 Molecular Formula: C8H10BNO3 Formula Weight: 178.98 Melting Point: 114-124° MDL No.: MFCD04038918

Vzorec: C₈H₁₀BNO₃
MW: 178,98 g/mol
Bod tání: 114…124 °C
Storage Temperature: Ambient
MDL Number: MFCD04038918
CAS číslo: 832695-88-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR