Tisk…

N,N-Dimethyl-1-dodecylamine N-oxide 0.1 mol/l vodný roztok, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: N-ethyl-N-oxido-dodecan-1-amine, LDAO, Lauryldimethylamine oxide, N,N-Dimethyldodecan-1-amine oxide, Lauramine oxide, N,N-Dimethyl-N-dodecylamine N-oxide

40231-250ML 40231-50ML
SIAL40231-250MLEA 0 CZK
SIAL40231-250ML SIAL40231-50ML
N,N-Dimethyl-1-dodecylamine N-oxide 0.1 mol/l vodný roztok, Sigma-Aldrich®
N,N-Dimethyl-1-dodecylamine N-oxide
Vzorec: C₁₄H₃₁NO
MW: 229,40 g/mol
MDL Number: MFCD00002049
CAS číslo: 1643-20-5
EINECS: 216-700-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR