Tisk…

Ethyl-2-methylthiazole-4-carboxylate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 2-methylthiazole-4-carboxylate

716308-1G 716308-5G
SIAL716308-1GEA 2240 CZK
SIAL716308-1G SIAL716308-5G
Ethyl-2-methylthiazole-4-carboxylate, Sigma-Aldrich®
Ethyl-2-methylthiazole-4-carboxylate
Vzorec: C₇H₉NO₂S
MW: 171,22 g/mol
MDL Number: MFCD00156148
CAS číslo: 6436-59-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR