5-(4-Formylphenyl)-1H-tetrazole ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-(1H-Tetrazol-5-yl)benzaldehyde
H50470.03
H50470.MD
H50470.03EA
9510
CZK
H50470.03
H50470.MD
5-(4-Formylphenyl)-1H-tetrazole ≥99%
5-(4-Formylphenyl)-1H-tetrazole
Purity: 99%
CAS No.: 74815-22-8
Molecular Formula: C8H6N4O
Formula Weight: 174.16
MDL No.: MFCD01311033
Vzorec:
C₈H₆N₄O MW: 174,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01311033 CAS číslo: 74815-22-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...