Tisk…

5-Oxazolecarboxylic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H30179.03 H30179.06
H30179.03EA 1610 CZK
H30179.03 H30179.06
5-Oxazolecarboxylic acid ≥98%
5-Oxazolecarboxylic acid

Purity: 98+% CAS No.: 118994-90-4 Molecular Formula: C4H3NO3 Formula Weight: 113.07 Melting Point: ca 245° dec. MDL No.: MFCD04114931

Vzorec: C₆H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD04114930
CAS číslo: 118994-89-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR