3-Amino-4-chloro-1H-indazole 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Chloro-1H-indazol-3-amine
H34143.MD
H34143.03
H34143.06
H34143.MDEA
1580
CZK
H34143.MD
H34143.03
H34143.06
3-Amino-4-chloro-1H-indazole 97%
3-Amino-4-chloro-1H-indazole
Vzorec:
C₇H₆ClN₃ MW: 167,60 g/mol Bod tání: 157…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00663139 CAS číslo: 20925-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...