Tisk…

1H-Indazole-5-boronic acid, Sigma-Aldrich®

Dodavatel: Merck

724017-250MG 724017-1G
SIAL724017-250MGEA 4170 CZK
SIAL724017-250MG SIAL724017-1G
1H-Indazole-5-boronic acid, Sigma-Aldrich®
1H-Indazole-5-boronic acid
Vzorec: C₇H₇BN₂O₂
MW: 161,9537 g/mol
MDL Number: MFCD06739151
CAS číslo: 338454-14-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR