Tisk…

Pyridazine-4-carboxylic acid ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Pyridazinecarboxylic acid

H27246.03 H27246.06
H27246.03EA 1290 CZK
H27246.03 H27246.06
Pyridazine-4-carboxylic acid ≥97%
Pyridazine-4-carboxylic acid

Purity: 97% CAS No.: 50681-25-9 Molecular Formula: C5H4N2O2 Formula Weight: 124.10 Melting Point: ca 244° dec. MDL No.: MFCD00011576

Vzorec: C₅H₄N₂O₂
MW: 124,10 g/mol
Bod tání: ∼ 244 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00011576
CAS číslo: 50681-25-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR