Pyridazine-3-carboxylic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
H34341.03
H34341.06
H34341.03EA
1970
CZK
H34341.03
H34341.06
Pyridazine-3-carboxylic acid ≥97%
Pyridazine-3-carboxylic acid
Vzorec:
C₅H₄N₂O₂ MW: 124,10 g/mol Bod tání: ∼ 195 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD01646333 CAS číslo: 2164-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...