4,5-Dibromo-3(2H)-pyridazinone ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4,5-Dibromopyridazin-3(2H)-one
A15994.06
A15994.14
A15994.06EA
3130
CZK
A15994.06
A15994.14
4,5-Dibromo-3(2H)-pyridazinone ≥98%
4,5-Dibromo-3(2H)-pyridazinone
Purity: 98%
CAS No.: 5788-58-9
Molecular Formula: C4H2Br2N2O
Formula Weight: 253.89
Melting Point: ca 220° dec.
Beilstein Reference: 121469
Vzorec:
C₄H₂Br₂N₂O MW: 253,89 g/mol Bod tání: ∼ 220 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00023641 CAS číslo: 5788-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...