Tisk…

6-Quinoxalinecarboxylic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Quinoxaline-6-carboxylic acid

M-Clarity™ quality level = MQ100

703834-1G 703834-5G
SIAL703834-1GEA 4790 CZK
SIAL703834-1G SIAL703834-5G
6-Quinoxalinecarboxylic acid, Sigma-Aldrich®
6-Quinoxalinecarboxylic acid
Vzorec: C₉H₆N₂O₂
MW: 174,16 g/mol
MDL Number: MFCD01365834
CAS číslo: 6925-00-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR