Tisk…

3-(tert-Butyldimethylsiloxy)thiophenol 95%

Dodavatel: Thermo Scientific
Varování

L17238.03 L17238.06
L17238.03EA 2340 CZK
L17238.03 L17238.06
3-(tert-Butyldimethylsiloxy)thiophenol 95%
3-(tert-Butyldimethylsiloxy)thiophenol

Purity: 95% CAS No.: 216393-56-5 Molecular Formula: C12H20OSSi Formula Weight: 240.44 Refractive Index: 1.5210

Vzorec: C₁₂H₂₀OSSi
MW: 240,44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318115
CAS číslo: 216393-56-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR