Tisk…

3-Ethynylthiophene, Sigma-Aldrich®

Dodavatel: Merck

578797-1G 578797-5G
SIAL578797-1GEA 3530 CZK
SIAL578797-1G SIAL578797-5G
3-Ethynylthiophene, Sigma-Aldrich®
3-Ethynylthiophene
Vzorec: C₆H₄S
MW: 108,16 g/mol
MDL Number: MFCD04039973
CAS číslo: 67237-53-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR