Tisk…

1-Methyl-1H-indazole-4-boronic acid hydrochloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (1-Methyl-1H-indazol-4-yl)boronic acid hydrochloride

723606-250MG 723606-1G
SIAL723606-250MGEA 4250 CZK
SIAL723606-250MG SIAL723606-1G
1-Methyl-1H-indazole-4-boronic acid hydrochloride, Sigma-Aldrich®
1-Methyl-1H-indazole-4-boronic acid hydrochloride
Vzorec: C₈H₁₀BClN₂O₂
MW: 212,4400 g/mol
MDL Number: MFCD13176537
CAS číslo: 1257527-53-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR