Dimethylzinc 2 mol/l in toluene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
364401-500ML
364401-50ML
SIAL364401-500MLEA
77080
CZK
SIAL364401-500ML
SIAL364401-50ML
Dimethylzinc 2 mol/l in toluene, Sigma-Aldrich®
Dimethylzinc
Vzorec:
C₂H₆Zn MW: 95.478 g/mol |
MDL Number:
MFCD00014854 CAS číslo: 544-97-8 EINECS: 208-884-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...