2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H50236.03
H50236.MD
H50236.03EA
4740
CZK
H50236.03
H50236.MD
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid ≥98%
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Purity: 98%
CAS No.: 123533-41-5
Molecular Formula: C14H10N2O2
Formula Weight: 238.25
MDL No.: MFCD00269226
Vzorec:
C₁₄H₁₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00269226 CAS číslo: 123533-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...