Tisk…

Ammonium sulphate-[¹⁵N₂] (98% ¹⁵N), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

299286-10G 299286-1G 299286-20G
SIAL299286-10GEA 32530 CZK
SIAL299286-10G SIAL299286-1G SIAL299286-20G
Ammonium sulphate-[¹⁵N₂] (98% ¹⁵N), Sigma-Aldrich®
Ammonium sulphate-[¹⁵N₂]
Vzorec: H₈N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00003392
CAS číslo: 43086-58-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR