Tisk…

Borane-tert-butylamine complex 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Boron hydride-tert-butylamine

L13177.09 L13177.18
L13177.09EA 1240 CZK
L13177.09 L13177.18
Borane-tert-butylamine complex 97%
Borane-tert-butylamine complex

Purity: 97% CAS No.: 7337-45-3 Molecular Formula: (CH3)3CNH2.BH3 Formula Weight: 86.98 Melting Point: 96-98° Beilstein Reference: 4134905 UN No.: 2811 MDL No.: MFCD00075635

Vzorec: C₄H₁₄BN
Bod tání: 96…98 °C
Hustota: 0,870
Storage Temperature: Ambient
MDL Number: MFCD00075635
CAS číslo: 7337-45-3
EINECS: 230-851-5
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR