4-Bromo-1H-pyrazole, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
374822-10G
374822-1G
SIAL374822-10GEA
0
CZK
SIAL374822-10G
SIAL374822-1G
4-Bromo-1H-pyrazole, Sigma-Aldrich®
4-Bromo-1H-pyrazole
Vzorec:
C₃H₃BrN₂ MW: 146,97 g/mol |
MDL Number:
MFCD00075602 CAS číslo: 2075-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...