Cyclobutylboronic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
646598-2G
646598-500MG
SIAL646598-2GEA
8050
CZK
SIAL646598-2G
SIAL646598-500MG
Cyclobutylboronic acid, Sigma-Aldrich®
Cyclobutylboronic acid
Vzorec:
C₄H₉BO₂ MW: 99.9241 g/mol |
MDL Number:
MFCD06798108 CAS číslo: 849052-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...