Tisk…

3-(2-Pyrrolidinyl)pyridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(3-Pyridyl)pyrrolidine, (±)-Nornicotine, DL-Nornicotine, 3-(Pyrrolidin-2-yl)pyridine

M-Clarity™ quality level = MQ200

N3018-25MG N3018-50MG
SIALN3018-25MGEA 6760 CZK
SIALN3018-25MG SIALN3018-50MG
3-(2-Pyrrolidinyl)pyridine, Sigma-Aldrich®
3-(2-Pyrrolidinyl)pyridine
Vzorec: C₉H₁₂N₂
MW: 148,21 g/mol
MDL Number: MFCD00016913
CAS číslo: 5746-86-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR