Tisk…
HPLC columns, Hypersil™ CPS and CPS-2 cyano
Dodavatel: Thermo Fisher Scientific
31805-254630 31805-154630 30805-154630 30805-254030 30805-124030 30803-154630 30803-104630 30805-254630
555-5260EA 23970 CZK
555-5260 555-5264 555-6719 555-6715 555-6724 555-6680 555-6684 555-6714
HPLC columns, Hypersil™ CPS and CPS-2 cyano
Kolony chromatografické
Hypersil™ CPS (cyano) phases offer different selectivity than Hypersil™ Silica and APS-2. Cyano columns equilibrate rapidly and are not deactivated by small quantities of water. When used as a reversed phase material, cyano selectivity complements the alkyl chain bonded phases, but with lower hydrophobicity. Hypersil™ CPS-2 is the endcapped version.
  • Operates in both normal and reversed phase modes
  • More rapid mobile phase equilibration compared with silica in normal phase
  • Not deactivated by traces of water in normal phase