Nitrification inhibitor for BOD
Dodavatel: Hach
253335
45901
253334
257924
662-0006EA
1890
CZK
662-0006
HACH45901
HACH253334
HACH257924
Nitrification inhibitor for BOD
Standardy
Standardy pro environmentální analýzu
TCMP - 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate.